Masterflex® Adapter Fittings, Sanitary Clamp to Male Thread, Straight, Avantor®
Supplier: Avantor Fluid Handling
40619-37 40619-42 40619-46 40619-49
MFLX31200-03EA 17765.94 INR
MFLX31200-03 MFLX31805-59 MFLX31805-63 MFLX31805-71 MFLX31201-02 MFLX31808-58 MFLX31808-60 MFLX31808-69 MFLX31808-77 MFLX31808-79 MFLX31808-80 MFLX40619-34 MFLX40619-35 MFLX40619-36 MFLX40619-38 MFLX40619-39 MFLX40619-40 MFLX40619-41 MFLX40619-44 MFLX40619-45 MFLX40619-47 MFLX40619-50 MFLX40619-51 MFLX40619-37 MFLX40619-42 MFLX40619-46 MFLX40619-49
Masterflex® Adapter Fittings, Sanitary Clamp to Male Thread, Straight, Avantor®
Fittings and Connectors
Maintain sanitary conditions while quickly and easily connecting to a male NPT thread.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for
More Product Information