Masterflex® Female Luer to Barb Fitting, Avantor®
Supplier: Avantor Fluid Handling
MFLX40314-00EA 930 INR
MFLX40314-00 MFLX40314-01 MFLX40314-02 MFLX40314-03 MFLX40314-04 MFLX40314-05 MFLX40314-06 MFLX40314-07 MFLX40314-08 MFLX40314-09 MFLX40314-10 MFLX40314-11 MFLX40314-12 MFLX40314-13 MFLX40314-14 MFLX40314-15 MFLX40314-16 MFLX40314-17 MFLX40314-18 MFLX40314-19 MFLX40314-20 MFLX40314-21 MFLX40314-22 MFLX40314-23 MFLX40314-24 MFLX40314-25 MFLX40314-26 MFLX40314-27 MFLX40314-28 MFLX40314-29 MFLX40314-30
Masterflex® Female Luer to Barb Fitting, Avantor®
Fittings and Connectors
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information