Masterflex® Barbed Micro2 Flow Valved Male Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
MFLX40262-37EA 0 INR
MFLX40262-37 MFLX40262-38 MFLX40262-39 MFLX40262-40 MFLX40262-41 MFLX40262-42 MFLX40262-43 MFLX40262-44 MFLX40262-45 MFLX40262-46 MFLX40262-47 MFLX40262-48 MFLX40262-49 MFLX40262-50 MFLX40262-51 MFLX40262-52
Masterflex® Barbed Micro2 Flow Valved Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination.
  • Aseptic connection and disconnection
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
  • A complete coupler requires a male and female fitting with same flow size (order separately).
  • Compatible with Female Micro2 Quick-Disconnect fittings.
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to passMore Product Information