Masterflex® Barbed High Flow Male Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
MFLX40250-64EA 0 INR
MFLX40250-64 MFLX40250-65 MFLX40250-66 MFLX40250-72 MFLX40250-73 MFLX40250-74 MFLX40250-80 MFLX40250-81 MFLX40250-82 MFLX40250-88 MFLX40250-89 MFLX40250-90 MFLX40250-95 MFLX40250-96 MFLX40250-97 MFLX40251-03 MFLX40251-04 MFLX40251-05
Masterflex® Barbed High Flow Male Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Female fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured,More Product Information