Masterflex® Barbed High Flow Female Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling
MFLX40250-04EA 7280 INR
MFLX40250-04 MFLX40250-05 MFLX40250-06 MFLX40250-12 MFLX40250-13 MFLX40250-14 MFLX40250-19 MFLX40250-20 MFLX40250-21 MFLX40250-27 MFLX40250-28 MFLX40250-29 MFLX40250-34 MFLX40250-35 MFLX40250-36 MFLX40250-42 MFLX40250-43 MFLX40250-44 MFLX40250-49 MFLX40250-50 MFLX40250-51 MFLX40250-57 MFLX40250-58 MFLX40250-59
Masterflex® Barbed High Flow Female Quick-Disconnect Fittings, Avantor®
Fittings and Connectors
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Male fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured, aMore Product Information