Masterflex® Direct-Read Variable-Area Flowmeter Kits with Interchangeable Scales, Avantor®
Supplier: Avantor Fluid Handling
34500-14 34500-18 34500-24 34500-28 34500-36 34500-44 34500-54
MFLX34500-10EA 14764.4 INR
MFLX34500-10 MFLX34500-12 MFLX34500-16 MFLX34500-22 MFLX34500-26 MFLX34500-30 MFLX34500-32 MFLX34500-34 MFLX34500-38 MFLX34500-40 MFLX34500-42 MFLX34500-46 MFLX34500-48 MFLX34500-52 MFLX34500-56 MFLX34500-58 MFLX34500-60 MFLX34500-14 MFLX34500-18 MFLX34500-24 MFLX34500-28 MFLX34500-36 MFLX34500-44 MFLX34500-54
Masterflex® Direct-Read Variable-Area Flowmeter Kits with Interchangeable Scales, Avantor®
Flow Meters
Seven interchangeable scales for routine gases and water offer flexibility.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Designed with stable, easy-to-read float for quick measurement readings
  • Interchangeable scales offer English and metric scales
  • Use with multiple routine gases for laboratory and industrial applications
  • Easy disassembly and assembly for cleaning
These easy-to-read acrylic flowmeter kits come with seven interchangeable back plates for routine gases- air, argon,More Product Information