Aalborg® Digital Gas Mass Flowmeters
Supplier: Avantor Fluid Handling
33400-04 33400-06 33400-08 33400-09 33400-27 33400-56 33400-57 33400-58 33401-06 33401-07 33401-08 33401-09 33401-19 33401-29 33401-52 33401-55 33403-44 33403-50 33403-77 33403-79 33400-36 33403-00 33403-55 33403-69
MFLX33400-04EA 177162.99 INR
MFLX33400-04 MFLX33400-06 MFLX33400-08 MFLX33400-09 MFLX33400-27 MFLX33400-56 MFLX33400-57 MFLX33400-58 MFLX33401-06 MFLX33401-07 MFLX33401-08 MFLX33401-09 MFLX33401-19 MFLX33401-29 MFLX33401-52 MFLX33401-55 MFLX33403-44 MFLX33403-50 MFLX33403-77 MFLX33403-79 MFLX33400-36 MFLX33403-00 MFLX33403-55 MFLX33403-69
Aalborg® Digital Gas Mass Flowmeters
Flow Meters
Stores calibration data for up to 10 gases.
  • Self-diagnostic tests
  • Automatic sensor zero offset adjustment
These digital flowmeters represent a new generation in mass flow technology. Meters incorporate a precision microcontroller and nonvolatile memory that stores all hardware specific variables, up to 10 different calibration tables, conversion factors for up to 32 gases, and supports up to 23 volumetric flow or mass flow engineering units, including user defined units. This feature allows the same meter to be calibrated for multiple gases whileMore Product Information