Chem-Tec Adjustable Standard Low-Flow Switiches
Supplier: Avantor Fluid Handling
32929-10 32929-16 32929-12 32929-14
MFLX32929-10EA 66000 INR
MFLX32929-10 MFLX32929-16 MFLX32929-12 MFLX32929-14
Chem-Tec Adjustable Standard Low-Flow Switiches
Flow Switches
Adjustable set points trigger SPST contacts rated for 70 VA.
  • Choose from brass or TFE materials
Standard Low-Flow Switches are normally closed (NC) but can be wired for normally open (NO). Typical applications include chemical process and vapor deposition systems; industrial gas lines; pollution control monitoring; atmospheric furnaces; and leak detection.
Choose from a brass switch (32929-10, -14) for pressures up to 1500 psi, or a switch made from PTFE (32929-12, -16) for good chemical compatibility. Wetted materials are brass, epoxy, and Viton forMore Product Information