Dwyer Sight Flow Indicators and Transmitters
Supplier: Avantor Fluid Handling
32779-44 32779-52
MFLX32779-44EA 12300 INR
MFLX32779-44 MFLX32779-52
Dwyer Sight Flow Indicators and Transmitters
Flow Indicators
Easily view flow indicator through clear plastic housing.
  • Models with 1 to 10 VDC output offer local and remote indication
  • No magnets in the rotor to attract ferrous materials
  • Output sensors can be added and replaced in the field without removing body from the process line
Sight flow indicators are a low-cost, durable rotor-style flow indicator. Choose from models that provide basic visual flow indication or models with output for remote flow monitoring. Units are constructed of clear plastic, enabling 360° viewing of the spinning rotor for easy flowMore Product Information