Masterflex® Thermal Mass Flowmeters/Flow Transmitters for Gases, Avantor®
Supplier: Avantor Fluid Handling
32707-14 32707-20 32707-22 32707-24 32707-26 32707-28 32707-30 32707-32 32707-36 32712-44 32707-00 32707-02 32707-04 32707-06 32707-08 32707-10 32707-12 32707-16 32707-34 32712-36 32712-40 32712-52
MFLX32707-14EA 85785.41 INR
MFLX32707-14 MFLX32707-20 MFLX32707-22 MFLX32707-24 MFLX32707-26 MFLX32707-28 MFLX32707-30 MFLX32707-32 MFLX32707-36 MFLX32712-44 MFLX32707-00 MFLX32707-02 MFLX32707-04 MFLX32707-06 MFLX32707-08 MFLX32707-10 MFLX32707-12 MFLX32707-16 MFLX32707-34 MFLX32712-36 MFLX32712-40 MFLX32712-52
Masterflex® Thermal Mass Flowmeters/Flow Transmitters for Gases, Avantor®
Flow Meters
Get faster performance with miniature coil design.
  • All meters feature 0 to 5 VDC output signal for data logging or controlling other instruments
These low-cost flowmeters use thermal gas flow sensing technique that results in highly accurate readings and repeatability. All units are calibrated to air/nitrogen—call our Application Specialists for meters calibrated to other gases or gas mixtures. Meters feature a cable hub connector for a quick connection to a power supply and 0 to 5 VDC output cable; order power supplies/cables under accessories.
More Product Information