Masterflex® Mass Flowmeter Controllers for Gases, Avantor®
Supplier: Avantor Fluid Handling
32708-02 32708-26
MFLX32708-02EA 242000 INR
MFLX32708-02 MFLX32708-26
Masterflex® Mass Flowmeter Controllers for Gases, Avantor®
Flow Switches
Integrated valve allows precise flow control.
  • Accepts a 0 to 5 VDC input signal for remote valve control
  • Calibrated for air and nitrogen
These low-cost mass flowmeter controllers use a thermal gas flow sensing technique that results in highly accurate readings and repeatability. The controllers accept a 0 to 5 VDC input signal for remote valve control. Controllers with LCD display feature a potentiometer for manual valve control. The 3 1/2"-digit display tilts up to 90° for easy viewing. Front panel dip switch lets you select one of the factoryMore Product Information