Kobold® Paddle Flow Switches, Polysulfone
Supplier: Avantor Fluid Handling
32481-27 32481-28
MFLX32481-27EA 34200 INR
MFLX32481-27 MFLX32481-28
Kobold® Paddle Flow Switches, Polysulfone
Flow Switches
Detect flow/no flow up to 225 °F (107 °C).
  • Easy to install
  • Low pressure drop
The Paddle Type Flow Switch is installable in tubes 1" and larger without the use of diaphragms, sealing bellows, or O-rings. These units, made of polysulfone, are almost maintenance free. The pressure drop across the instrument is less than 1.45PSI and independent of the pipe diameter. Operation of the unit is clearly visible through the polysulfone housing. They are available with either a normally open or a normally closed contact. Installation is extraordinarily simple.More Product Information