Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters, 65-mm Scale, Avantor®
Supplier: Avantor Fluid Handling
32010-01 32010-09 32010-11 32010-15 32010-16 32010-31 32010-35 32011-03 32011-07 32011-09 32011-13 32011-16 32011-17 32011-31 32011-33 32012-01 32012-05 32012-09 32012-15 32012-16 32012-29 32012-31 32012-35
MFLX32010-03EA 21418.42 INR
MFLX32010-03 MFLX32010-05 MFLX32010-07 MFLX32010-13 MFLX32010-17 MFLX32011-01 MFLX32012-03 MFLX32012-07 MFLX32012-11 MFLX32012-13 MFLX32012-17 MFLX32010-01 MFLX32010-09 MFLX32010-11 MFLX32010-15 MFLX32010-16 MFLX32010-31 MFLX32010-35 MFLX32011-03 MFLX32011-07 MFLX32011-09 MFLX32011-13 MFLX32011-16 MFLX32011-17 MFLX32011-31 MFLX32011-33 MFLX32012-01 MFLX32012-05 MFLX32012-09 MFLX32012-15 MFLX32012-16 MFLX32012-29 MFLX32012-31 MFLX32012-35
Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters, 65-mm Scale, Avantor®
Flow Meters
No more eye strain - front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
More Product Information