CPC® Miniature Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Supplier: Avantor Fluid Handling
31604-61
MFLX31604-61EA 5900 INR
MFLX31604-61 MFLX31604-54 MFLX31604-57
CPC® Miniature Non-Spill Quick-Disconnect Fittings, Bodies and Inserts
Fittings and Connectors
Stop leaks in your line—spillage as low as 0.025mL.
  • Non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety
  • Easy one-hand operation
These CPC® non-spill quick-disconnect fittings virtually eliminate any loss of product with this leak-free design. Couplings are made of polypropylene with an EPDM O-ring for excellent chemical resistance. These non-spill series of couplings ensure minimal leaks and air inclusions in your process line—protects process purity and minimizes loss due to accidental disconnects. The NS2More Product Information