Sanitary Tri-Clamp End Caps, Stainless Steel
Supplier: Avantor Fluid Handling
MFLX30508-11EA 7240.43 INR
MFLX30508-11
Sanitary Tri-Clamp End Caps, Stainless Steel
Fittings and Connectors
Sanitary seals open end to prevent debris from accumulating.
  • Ouick and easy to mount or dismount with a pressure clamp
The tri-clamp end caps play an important role in the sanitation process, allowing an open end to be sanitary seals preventing additional debris from accumulating.