Hayward® Simplex Basket Strainers, PVC
Supplier: Avantor Fluid Handling
29526-06 29526-10 29526-12
MFLX29526-06EA 185000 INR
MFLX29526-06 MFLX29526-10 MFLX29526-12
Hayward® Simplex Basket Strainers, PVC
Filters Strainers
Ideal product for the removal of crude solids.
  • Low pressure drop with large straining capacity
  • Basket easily lifts out to clean or replace
  • True union connections permit easy removal of strainer assesmbly from process line
Protect pipes and process equipment from dirt and debris. The perforated baskets catch unwanted particles as the fluid passes through the strainer. Suitable for use in industries processing water, oil, paint, chemicals, or food.
Strainers are made of PVC and include FPM seals. The 1⁄2", 3⁄4", and 1" strainers include a 1⁄32"More Product Information