Masterflex® Single-Use Turbine Flow Sensor Bundles, Avantor®
Supplier: Avantor Fluid Handling
MFLX24507-80EA 232700 INR
MFLX24507-80
Masterflex® Single-Use Turbine Flow Sensor Bundles, Avantor®
Fluid Sensors
Ready-to-go flow sensor system bundles for your application.
  • Order a bundle for easy start up - flow sensor and tubes bundled together
  • Modular interchangeable flow sensor systems with single-use design
  • Accurate 1% of reading measurement at low flow
  • Low operating cost disposable flow tubes for optimal contamination-free production
  • Ideal for single-use applications while also suitable for long-term measurement
  • Flow tubes and turbine sensors are produced and tested under cleanroom conditions
These Masterflex® single-use liquid turbine flow sensors areMore Product Information