Argos Technologies® Flexifuge Microcentrifuge
Supplier: Avantor Fluid Handling
22999-50 22999-51
MFLX22999-50EA 0 INR
MFLX22999-50 MFLX22999-51
Argos Technologies® Flexifuge Microcentrifuge
Centrifuges Microcentrifuges
Change color of illuminated LED light ring to personalize sample spinning.
  • Quick centrifugation in a tiny footprint
  • Versatile rotor options suit many standard lab needs including 5 ml microtubes
  • Operate in continuous, pulsed, or timed mode
This compact versatile benchtop centrifuge is ideal for quick two-speed spin downs. Three rotors accommodate PCR, 0.5 ml, 1.5 ml, and 5 ml tubes. Operate the centrifuge in continuous mode, pulsed for a short burst of rapid centrifugation, or timed to complete the spin in 1 to 15 minutes.
Regardless of setting, rotorMore Product Information