PendoTECH® Single-Use Pressure Sensors for Pressure Monitor and Transmitter Systems
Supplier: Avantor Fluid Handling
MFLX19406-20EA 0 INR
MFLX19406-20 MFLX19406-21 MFLX19406-22 MFLX19406-23 MFLX19406-24 MFLX19406-25 MFLX19406-27 MFLX19406-28 MFLX19406-30 MFLX19406-31 MFLX19406-32 MFLX19406-51 MFLX19406-52 MFLX19406-53 MFLX19406-54 MFLX19406-55 MFLX19406-56 MFLX19406-29
PendoTECH® Single-Use Pressure Sensors for Pressure Monitor and Transmitter Systems
Fluid Sensors
Single-use solution to monitor process pressure.
  • Ideal for applications such as tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring, and chromatography
  • Sensors priced for single-use systems, but may be reused
Sensors measure static and dynamic process pressure and are perfect components for disposable systems requiring sterilization, but also minimal downtime. Sensors feature an integral flow sensor with no dead legs and no fluid path obstructions. All pressure sensors are manufactured in a FDA compliant facility,More Product Information