BIOLiner High Capacity Fixed Angle Rotor, Buckets, and Adapters
Supplier: Avantor Fluid Handling
17708-46 17708-47 17708-51 17708-40 17708-41 17708-42
MFLX17708-46EA 0 INR
MFLX17708-46 MFLX17708-47 MFLX17708-51 MFLX17708-40 MFLX17708-41 MFLX17708-42
BIOLiner High Capacity Fixed Angle Rotor, Buckets, and Adapters
Shakers Shaking Incubators
High capacity rotor provides versatility and optional biocontainment capabilities.
The BIOLiner high capacity rotor provides versatility to your system and provides protection from samples with the optional biocontainment lids. Lids snap easily in place with just one hand. Three easy steps to change your configuration:
- Release rotor by pressing button located in the center of the rotor.
- Remove and attach new buckets/adapters by snapping into place.
- Press rotor back in place until it is secure.
Adapters provide you additional versatility by accomodating sample containers from 5 mlMore Product Information