Thermo Scientific Megafuge 8 Centrifuge
Supplier: Avantor Fluid Handling
17701-20
MFLX17701-20EA 0 INR
MFLX17701-20
Thermo Scientific Megafuge 8 Centrifuge
Centrifuges Benchtop Centrifuges
Exceptional capacity and flexible processing adds value to any lab.
  • SMARTSpin imbalance detection system prevents hazardous sample running
  • Quality design and brushless drive ensure years of quiet use
  • Biocontainment option keeps you safe
Conforming to the latest safety standards, the Megafuge 8 is a compact bench centrifuge that satisfies many sample running options. The secure push-button Auto-Lock® rotor exchange system with glove-friendly design allows you to change out rotors with ease, requiring only one hand! Choose from several rotor types,More Product Information