Masterflex® MPR Series Multipurpose Refrigerated Centrifuges, Avantor®
Supplier: Avantor Fluid Handling
17406-05 17406-15 17406-16 17406-18 17406-19 17406-20 17406-00 17406-10 17406-25
MFLX17406-05EA 0 INR
MFLX17406-05 MFLX17406-15 MFLX17406-16 MFLX17406-18 MFLX17406-19 MFLX17406-20 MFLX17406-00 MFLX17406-10 MFLX17406-25
Masterflex® MPR Series Multipurpose Refrigerated Centrifuges, Avantor®
Centrifuges Benchtop Centrifuges
Rugged construction without sacrificing reliable performance.
  • Extra-thick stainless steel chamber and zinc-coated alloy and steel frame increase durability for inorganic separations requiring strong solvents
  • Industrial-grade inverter drive (VFD) and brushless motor provide a long service life
  • User-accessible service sections allow on-site maintenance
  • Programmable memory quickly administers routine tests and accommodates multiple users
  • Pulse feature lets you perform very short runs to 90 seconds, perfect for fast pelleting
This Cole-Parmer multipurposeMore Product Information