Koehler Portable Automatic Heated Oil Test Centrifuges
Supplier: Avantor Fluid Handling
17301-09 17301-11
MFLX17301-09EA 0 INR
MFLX17301-09 MFLX17301-11
Koehler Portable Automatic Heated Oil Test Centrifuges
Centrifuges Benchtop Centrifuges
Integrated preheating chamber for convenient preconditioning of your samples.
  • Conforms to the specifications of ASTM D96 and API MPMS Chapter 10.4
  • Independent digital temperature displays for both the preheating chamber and rotor bowl
  • Thoroughly insulated to prevent heat loss in your sample
Use this fully portable oil test centrifuge for water and sediment determination in crude oil during field custody transfers. The unit accommodates two conical short tubes (order separately). An integral four-place tube chamber allows you to set up two tubes, thenMore Product Information