Masterflex® Transfer PTFE Hose Assemblies, Avantor®
Supplier: Avantor Fluid Handling
16382-10
MFLX16382-01EA 16700 INR
MFLX16382-01 MFLX16382-04 MFLX16382-09 MFLX16382-18 MFLX16383-15 MFLX16382-10
Masterflex® Transfer PTFE Hose Assemblies, Avantor®
Tubing Transfer Tubing
Custom-made transfer hose assemblies feature sanitary Tri-Clamp® end fittings.
  • Exceptional resistance to mechanical stress, high temperatures and oxidation
  • Co-extruded clear/white pigmented, phthalate-free, smooth, tested hose
  • Reinforcement of stainless-steel wire helices and synthetic plies
  • Smooth, white and glossy silicone cover which is heat, abrasion, aging and ozone resistant
  • Hose in compliance with 1907/2006/CE(REACH) meeting FDA 21 CFR 177.1550 standards and USP Class VI requirements
  • Test certificates and documentation for 100% quantity available
More Product Information