Value Plastics Large-Bore Barbed Connectors 65389
Supplier: Avantor Fluid Handling
15955-68 15955-73 15955-55 15955-67 15955-62 15955-70 15955-58 15955-65
MFLX15955-68EA 9700 INR
MFLX15955-68 MFLX15955-73 MFLX15955-55 MFLX15955-67 MFLX15955-62 MFLX15955-70 MFLX15955-58 MFLX15955-65
Value Plastics Large-Bore Barbed Connectors 65389
Fittings and Connectors
Top-quality fittings from 100% virgin materials.
  • Manufactured in an ISO 13533-1 Class 8 (formerly Class 100,000) cleanroom
  • Full material traceability on all items
Use these premium barbed connectors with any flexible tubing in a fluid handling line. Manufactured to strict quality standards under an ISO 9001:2000 quality control system, these fittings from Value Plastics meet your needs with quality and low cost time after time.
Generally, barbed fittings are a sensible choice with flexible tubing in low-pressure applications (typically less than 2.1More Product Information