Masterflex® Adapter Fittings, Hose Barb, Avantor®
Supplier: Avantor Fluid Handling
15955-04 15955-31 15955-36 15955-15 15955-26 15954-97 15955-40 15954-91 15955-24 15955-29 15955-41 15954-96 15955-07 15955-08 15955-06 15955-03
MFLX15955-04EA 5600 INR
MFLX15955-04 MFLX15955-31 MFLX15955-36 MFLX15955-15 MFLX15955-26 MFLX15954-97 MFLX15955-40 MFLX15954-91 MFLX15955-24 MFLX15955-29 MFLX15955-41 MFLX15954-96 MFLX15955-07 MFLX15955-08 MFLX15955-06 MFLX15955-03
Masterflex® Adapter Fittings, Hose Barb, Avantor®
Fittings and Connectors
Premium craftsmanship and 100% virgin materials produce the best quality fittings.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for
More Product Information