Masterflex® L/S® Multichannel Cartridge Pump Head with Reduced Pulsation, 12-Channel, Avantor®
Supplier: Avantor Fluid Handling
MFLX07519-25EA 179194.71 INR
MFLX07519-25
Masterflex® L/S® Multichannel Cartridge Pump Head with Reduced Pulsation, 12-Channel, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Synchronous, low-pulsation flow from multiple channels.
  • Cartridges snap in and out for fast tubing changes—no need to remove pump head from the drive
  • Improved retainer design holds tubing securely in place
  • Flow rate 0.00001 to 41 ml/min with microbore 2-stop tubing—flow rate depends on drive rpm, cartridge, and tubing size
  • Accepts up to 12 large cartridges
  • Require drives with a minimum of 1⁄10 hp and a maximum of 600 rpm
  • Constructed with 8 Rulon® rollers, stainless steel rotor, and PSF ends
Each pump head system is composed of a heavy-duty pump head,More Product Information