Submersible Pump Float Switch with Piggyback Plug
Supplier: Avantor Fluid Handling
07187-40
MFLX07187-40EA 14900 INR
MFLX07187-40
Submersible Pump Float Switch with Piggyback Plug
Level Sensors
Convert any manually controlled pump up to 1/2 hp into automatic operation.
  • Constructed of PVC and neoprene
Use this float switch to control submersible pumps that don''t have any automatic control. Simply plug the pump''s power cord into the back of the float switch plug (piggyback) and insert the float switch plug into a power outlet. The float switch will control the power to the pump based on the position of the float. Note: Must be used with pumps with a three-prong grounded power plug.
Delivery information: Includes 10-ft (3-m) cord with piggyback plug.