Masterflex® L/S® Standard Long-Shaft Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
MFLX07013-10EA 35282.02 INR
MFLX07013-10 MFLX07015-00 MFLX07015-10 MFLX07015-50 MFLX07016-00 MFLX07016-10 MFLX07024-00 MFLX07024-10
Masterflex® L/S® Standard Long-Shaft Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
This Standard pump head accepts a single tubing size to ensure optimal performance from your Masterflex pump. Use with Masterflex L/S pump drives requiring the long-shaft style pump head. These pump heads areMore Product Information