Masterflex® Transfer Tubing, PVC Reinforced, Avantor®
Supplier: Avantor Fluid Handling
06602-10 06602-12 06602-14 06602-16 06602-18 06602-20 06602-22 06602-26 06602-24
MFLX06602-10EA 26737.66 INR
MFLX06602-10 MFLX06602-12 MFLX06602-14 MFLX06602-16 MFLX06602-18 MFLX06602-20 MFLX06602-22 MFLX06602-26 MFLX06602-24
Masterflex® Transfer Tubing, PVC Reinforced, Avantor®
Tubing Transfer Tubing
More durable than standard PVC.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: High-pressure use, food processing, and beverage handling.
Characteristics: Good acid/alkali resistance and aging characteristics. White polyester yarn with orange tracer yarn embedded in a transparent PVC wall.
More Product Information