Masterflex® Ismatec® Microbore Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Supplier: Avantor Fluid Handling
MFLX06449-10EA 30465.78 INR
MFLX06449-10 MFLX06449-12 MFLX06449-14 MFLX06449-18 MFLX06449-22 MFLX06449-24 MFLX06449-26 MFLX06449-28 MFLX06449-30 MFLX06449-34 MFLX06449-36 MFLX06449-40 MFLX06449-42 MFLX06449-48 MFLX06449-31 MFLX06449-33 MFLX06449-37 MFLX06449-47
Masterflex® Ismatec® Microbore Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Tubing Peristaltic Pump Tubing
Longest life of all Tygon® formulations.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Applications: General laboratory use
Characteristics: Long life, won’t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic,More Product Information