Masterflex® L/S® Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Supplier: Avantor Fluid Handling
MFLX06440-13EA 23700 INR
MFLX06440-13 MFLX06440-14 MFLX06440-16 MFLX06440-17 MFLX06440-18 MFLX06440-25
Masterflex® L/S® Precision Pump Tubing, Tygon® S3™ E-LFL, Avantor®
Tubing Peristaltic Pump Tubing
Longest life of all Tygon® formulations.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-
More Product Information