Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, Avantor®
Supplier: Avantor Fluid Handling
MFLX06402-15EA 33262.41 INR
MFLX06402-15 MFLX06402-24 MFLX06402-35 MFLX06402-36
Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, Avantor®
Tubing Peristaltic Pump Tubing
Similar to Norprene® but with FDA approval.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-
More Product Information