Thermo Scientific Nalgene Centrifuge Bottles, 450- and 500-mL
Supplier: Avantor Fluid Handling
06105-55 06106-30 06106-35 06106-25
MFLX06105-55EA 0 INR
MFLX06105-55 MFLX06106-30 MFLX06106-35 MFLX06106-25
Thermo Scientific Nalgene Centrifuge Bottles, 450- and 500-mL
Bottles Centrifuge Bottles
Get better separation of large samples.
  • Ideal for low-speed centrifugation of biological and chemical samples
  • Easy view liquids
  • Autoclavable
These centrifuge bottles are available in polypropylene copolymer (PPCO) and polycarbonate (PC).
The PPCO bottles allow you to perform biopharmaceutical aseptic fluid transfer or sampling applications. Molded of transparent polycarbonate, these bottles are ideal for arge-volume cell harvesting, pelleting and protein purification.
The PC rounded bottom bottles are ideal for processing large-volume cell harvesting,More Product Information