Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Supplier: Avantor Fluid Handling
03229-35 03295-32 03295-34 03295-36
MFLX03217-78EA 75800 INR
MFLX03217-78 MFLX03227-32 MFLX03227-34 MFLX03227-36 MFLX03227-87 MFLX03229-33 MFLX03229-37 MFLX03229-35 MFLX03295-32 MFLX03295-34 MFLX03295-36
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 150-mm, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet,More Product Information