Eppendorf 5810 Series Multi-Purpose Non-Refrigerated Centrifuges
Supplier: Avantor Fluid Handling
02570-01
MFLX02570-01EA 0 INR
MFLX02570-01
Eppendorf 5810 Series Multi-Purpose Non-Refrigerated Centrifuges
Centrifuges Benchtop Centrifuges
Versatile rotor and adapter options can be used for virtually any application!
  • Superior quality and metal safety containment construction with aerosol tight lid options ensures your safety
  • Software control guarantees high accuracy, speed and temperature for optimal performance
Use these high-speed, multi-purpose centrifuges for a variety of applications: cell harvesting, high-speed separation of cell lysates, and DNA precipitations. The automatic rotor sensing feature will recognize your rotors and detect imbalances for safe operation. Order rotorsMore Product Information