Masterflex® PVC Pressure Relief Valves, Avantor®
Supplier: Avantor Fluid Handling
MFLX00918-50EA 52400 INR
MFLX00918-50
Masterflex® PVC Pressure Relief Valves, Avantor®
Valves Pressure-Relief Valves
Easily meter fluids with pressure relief valves.
  • Choose from different max pressures based on your application
These PVC valves have O-rings of Viton®, making them Ideal for liquid applications.