Poloxamer, Corning®
Supplier: Corning
61-161-RM 13-901-CI
MDTC61-161-RMEA 20161.86 INR
MDTC61-161-RM MDTC13-901-CI
Poloxamer, Corning®
Cell Culture Media
Poloxamer 188 is a surfactant used in cell culture to protect cells in suspension against possible damage during transfer, freezing and thawing, and stirring.
It is a non-ionic detergent with an average molecular weight of 8400Da consisting of a central polypropylene flanked by two polyethylene molecules.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature Refrigerate