Cavity slides for diagnosis
Supplier: MARIENFELD
1216520
MARI1216520EA 0 INR
MARI1216520
Cavity slides for diagnosis
Slides Cavity Slides
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
  • Soda-lime glass with epoxy resin colour masks
  • High resistance against solvents