Phenol solution_SC, StandARd®, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Warning

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

M.O.C. Category N

MACRH302-01EA 0 INR
MACRH302-01
Phenol solution_SC, StandARd®, Macron Fine Chemicals™
Phenol
Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Flash Pt: 81
MDL Number: MFCD00002143
CAS Number: 108-95-2
UN: 1671
ADR: 6.1 II
Merck Index: 12,07390

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Concentration (0.8-1.2 mg/mL) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR