Iodine 1.0 N, StandARd® Volumetric, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger


M.O.C. Category N

MACRH185-05EA 0 INR
MACRH185-05
Iodine 1.0 N, StandARd® Volumetric, Macron Fine Chemicals™
Iodine

Contains 10 to 20% iodine and 20 to 30% potassium iodide.

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8, 6.1 III
Merck Index: 13,05036

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.95 - 1.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR