Calcium sulphate, Drierite™ (8 Mesh), Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Danger

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum

MACR3378-04EA 0 INR
MACR3378-04
Calcium sulphate, Drierite™ (8 Mesh), Macron Fine Chemicals™
Calcium sulphate
Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1350
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
UN: 3077
ADR: 9 III
Merck Index: 13,01711

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR