VWR®RapidBlock™, Blocking solution, 10x
Supplier: VWR Chemicals
M325-100MLEA 0 INR
M325-100ML
VWR®RapidBlock™, Blocking solution, 10x
Buffers Electrophoresis Buffers Blocking Buffers
RapidBlock™ solution reduces blocking time to five minutes for Western and dot blotting procedures on PVDF and nitrocellulose membranes. This protein-free formulation minimises cross-reactivity and non specific antibody binding, generating blots with low background and enhanced signal-to-noise ratio.
  • Only 5 minutes for PVDF or nitrocellulose blocking
  • Reduce background and cross-reactivity
  • Enhance chemiluminescent signals
  • Protein-free formulation