Sodium dodecyl sulphate (SDS) 100 g/l aqueous solution, Proteomics grade

Supplier: VWR Chemicals
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

M111-100MLEA 0 INR
M111-100ML
Sodium dodecyl sulphate (SDS) 100 g/l aqueous solution, Proteomics grade
Sodium dodecyl sulphate (SDS)

Strong detergent utilised in lysis buffers for the complete disruption of membranes and denaturation of proteins.


  • High purity and protease-free
  • Suitable for most biological applications
  • Used in denaturing gel electrophoresis

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Colour (APHA) 5
DNase NONE
Protease NONE
RNase NONE


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR