Perfection and Performance Laboratory Underbench Freezers, SFTvh and SFTsg
Supplier: Liebherr
SFTVH1501 SFTSG1501 SFTSG1501_740
LIEBSFTVH1501EA 0 INR
LIEBSFTVH1501 LIEBSFTSG1501 LIEBSFTSG1501_740
Perfection and Performance Laboratory Underbench Freezers, SFTvh and SFTsg
Freezers Laboratory / General Purpose Freezers
Underbench freezers with forced air or static cooling. White steel exterior and commercial-grade polystyrol interior. The intuitive user interfaces provide useful extended multi-functionality, e.g. a maintenance timer. Advanced connectivity options to suit varying requirements for simplified data storage, temperature control and alarm signals.
  • Extended range of models for more choice to fulfil specific user requirements
  • Hygienic and long-lasting seamless polystyrol interior with a 10 mm access port for external probes
  • Energy-efficiency optimised appliances
More Product Information