Serological pipettes, with plugging top, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
37034A-10
KIML37034A-10EA 0 INR
KIML37034A-10
Serological pipettes, with plugging top, KIMAX®, KIMBLE®
Pipettes Serological Pipettes
Borosilicate glass, reusable.
  • Scale is permanent brown stain fused into the surface of uniform bore tubing without etching
  • Plain, fire-polished top end has been tooled to accept cotton plugging
  • Colour-coding facilitates sorting and selection
These KIMAX® pipettes are intended for bacteriological, microbiological, or serological work. Pipettes must be blown out after free delivery to obtain the total rated capacity.
Certifications: Designed from ASTM Specification E1044, Style II requirements