Filtering flasks, with hose barb connector, KIMBLE®
Supplier: KIMBLEKONTES
27070-4000
KIML27070-4000EA 0 INR
KIML27070-4000
Filtering flasks, with hose barb connector, KIMBLE®
Flasks Filter Flasks
Manufactured from 33 expansion, low extractable borosilicate glass, KIMAX®.
  • With a heavy wall
  • Designed for vacuum to 29" of mercury
  • Proper positioning of the connector provides greater flask stability
These flasks have a quick-release connector designed as a safety feature. And the hose barb connector can accommodate tubing of int.Ø ¼ inch and this tubing can remain permanently attached.