Bed pan
Supplier: KARTELL
586
KART586EA 0 INR
KART586
Bed pan
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Bed pan moulded in white PP.
  • Withstand temperatures up to 120 °C
  • Very good chemical resistance
  • Easy to clean