VWR® Buffer, MES-SDS, 20X, UltraPure Grade
Supplier: VWR Chemicals
K856-500MLEA 0 INR
K856-500ML
VWR® Buffer, MES-SDS, 20X, UltraPure Grade
Buffers Electrophoresis Buffers Running Buffers
Useful for high resolution of proteins on neutral pH SDS-PAGE gels.
  • Ultra-pure grade
  • Convenient concentrations and packaging
  • Good quality, consistency and resolution