Phenol : chloroform : iso-amyl alcohol (25:24:1) v/v/v 99,0, Biotechnology Grade for biotechnology 1 Phase

Supplier: VWR Chemicals
Danger

Synonyms: Carbolic acid, Hydroxybenzene, Phenic acid

K169-400MLEA 0 INR
K169-400ML
Phenol : chloroform : iso-amyl alcohol (25:24:1) v/v/v 99,0, Biotechnology Grade for biotechnology 1 Phase
Phenol : Chloroform : iso-Amyl alcohol

Ready-to-use, single-phase solution that does not have to be equilibrated with a buffering purification solution prior to use. Premixed with Isoamyl Alcohol (25:24:1).

Storage Temperature: Ambient
MDL Number: MFCD00133763
CAS Number: 136112-00-0
UN: 2810
ADR: 6.1 II

Order Now

Specification Test Results

Conductivity(50%, Water) @25 °C 250 uS - 330 uS
pH(50%, Water) @25°C 8.05 - 8.35


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR